(S)-Methyl 1-Tritylaziridine-2-carboxylate 97%

BD217355-1G BD217355-250MG
BLDPBD217355-1GEA 10.5 EUR
BLDPBD217355-1G BLDPBD217355-250MG
(S)-Methyl 1-Tritylaziridine-2-carboxylate 97%
(S)-Methyl 1-Tritylaziridine-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₂₃H₂₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD01863736
CAS Number: 75154-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR