2,6-Dibromophenol 98%
|
|
BD21742-100G
BD21742-1000G
BLDPBD21742-100GEA
144
EUR
BLDPBD21742-100G
BLDPBD21742-1000G
2,6-Dibromophenol 98%
2,6-Dibromophenol
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₆H₄Br₂O Storage Temperature: Ambient |
MDL Number:
MFCD00002152 CAS Number: 608-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...