(S)-Morpholin-3-ylmethanol hydrochloride 95%

BD217701-5G BD217701-100MG
BLDPBD217701-5GEA 373 EUR
BLDPBD217701-5G BLDPBD217701-100MG
(S)-Morpholin-3-ylmethanol hydrochloride 95%
(S)-Morpholin-3-ylmethanol hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₁₂ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12547468
CAS Number: 218594-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR