4-Bromo-2,3-dimethylpyridine 98%
|
|
BD218291-1G
BD218291-250MG
BLDPBD218291-1GEA
85.1
EUR
BLDPBD218291-1G
BLDPBD218291-250MG
4-Bromo-2,3-dimethylpyridine 98%
4-Bromo-2,3-dimethylpyridine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₇H₈BrN Storage Temperature: Refrigerator |
MDL Number:
MFCD18253389 CAS Number: 259807-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...