Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 98%

BD21851-25G BD21851-1000G
BLDPBD21851-25GEA 7.4 EUR
BLDPBD21851-25G BLDPBD21851-1000G
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 98%
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl)

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00012871
CAS Number: 623-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR