cis-4-Aminocyclohexanol hydrochloride 97%
|
|
BD218880-5G
BD218880-250MG
BD218880-1G
BLDPBD218880-5GEA
47.3
EUR
BLDPBD218880-5G
BLDPBD218880-250MG
BLDPBD218880-1G
cis-4-Aminocyclohexanol hydrochloride 97%
cis-4-Aminocyclohexanol hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₁₄ClNO Storage Temperature: Refrigerator |
MDL Number:
MFCD07366531 CAS Number: 56239-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...