1,4-Bis(2-benzoxazolyl)naphthalene 98%

BD21921-10G BD21921-5G BD21921-25G
BLDPBD21921-10GEA 10.5 EUR
BLDPBD21921-10G BLDPBD21921-5G BLDPBD21921-25G
1,4-Bis(2-benzoxazolyl)naphthalene 98%
1,4-Bis(2-benzoxazolyl)naphthalene

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₄H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00357168
CAS Number: 5089-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR