Methyl 2-(piperidin-1-ylmethyl)benzoate 98+%

BD219547-1G BD219547-5G
BLDPBD219547-1GEA 75.6 EUR
BLDPBD219547-1G BLDPBD219547-5G
Methyl 2-(piperidin-1-ylmethyl)benzoate 98+%
Methyl 2-(piperidin-1-ylmethyl)benzoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₁₉NO₂
MW: 233.30 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08272686
CAS Number: 914347-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR