2-Methoxy-5-nitrophenol 98%

BD21955-25G BD21955-1000G
BLDPBD21955-25GEA 16.8 EUR
BLDPBD21955-25G BLDPBD21955-1000G
2-Methoxy-5-nitrophenol 98%
2-Methoxy-5-nitrophenol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₇NO₄
MW: 169.14 g/mol
Boiling Pt: 320
Storage Temperature: Ambient
MDL Number: MFCD00015561
CAS Number: 636-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR