trans-3-Aminocyclobutanol hydrochloride 95%

BD219890-10G BD219890-250MG
BLDPBD219890-10GEA 748 EUR
BLDPBD219890-10G BLDPBD219890-250MG
trans-3-Aminocyclobutanol hydrochloride 95%
trans-3-Aminocyclobutanol hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD17214232
CAS Number: 1205037-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR