(S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate hydrochloride 95%
BD220177-100MG
BD220177-250MG
BLDPBD220177-100MGEA
30.5
EUR
BLDPBD220177-100MG
BLDPBD220177-250MG
(S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate hydrochloride 95%
(S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate hydrochloride
Formula:
C₁₀H₂₁ClN₂O₂ |
MDL Number:
MFCD11101393 CAS Number: 1070968-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...