4-Bromo-2-iodo-1-(trifluoromethyl)benzene 95%

BD220239-1G BD220239-10G
BLDPBD220239-1GEA 23.1 EUR
BLDPBD220239-1G BLDPBD220239-10G
4-Bromo-2-iodo-1-(trifluoromethyl)benzene 95%
4-Bromo-2-iodo-1-(trifluoromethyl)benzene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₇H₃BrF₃I
Storage Temperature: Refrigerator
MDL Number: MFCD21603817
CAS Number: 1256945-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR