N-Boc-N,N'-dimethylethylamine 97%

BD220403-1G BD220403-100G
BLDPBD220403-1GEA 11.6 EUR
BLDPBD220403-1G BLDPBD220403-100G
N-Boc-N,N'-dimethylethylamine 97%
N-Boc-N,N'-dimethylethylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₂₀N₂O₂
MW: 188.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953267
CAS Number: 112257-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR