7-Fluoro-3,4-dihydroquinolin-2(1H)-one 98%

BD220740-1G BD220740-100MG
BLDPBD220740-1GEA 361 EUR
BLDPBD220740-1G BLDPBD220740-100MG
7-Fluoro-3,4-dihydroquinolin-2(1H)-one 98%
7-Fluoro-3,4-dihydroquinolin-2(1H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD11811768
CAS Number: 4590-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR