3-Chloro-6-(trifluoromethyl)pyridin-2-amine 96%

BD221303-1G BD221303-250MG
BLDPBD221303-1GEA 472 EUR
BLDPBD221303-1G BLDPBD221303-250MG
3-Chloro-6-(trifluoromethyl)pyridin-2-amine 96%
3-Chloro-6-(trifluoromethyl)pyridin-2-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₄ClF₃N₂
MW: 196.55 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04037237
CAS Number: 886762-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR