Picolinic acid hydrochloride 98%
|
|
BD222099-5G
BD222099-100G
BLDPBD222099-5GEA
22.1
EUR
BLDPBD222099-5G
BLDPBD222099-100G
Picolinic acid hydrochloride 98%
Picolinic acid hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₆H₆ClNO₂ MW: 159.5703 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00050666 CAS Number: 636-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...