1-Methyl-1H-indazole-6-carboxylic acid 98%

BD222155-100MG BD222155-250MG
BLDPBD222155-100MGEA 17.9 EUR
BLDPBD222155-100MG BLDPBD222155-250MG
1-Methyl-1H-indazole-6-carboxylic acid 98%
1-Methyl-1H-indazole-6-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11043008
CAS Number: 1031417-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR