7-Bromoquinoline-2-carboxylic acid 97%

BD222173-5G BD222173-10G
BLDPBD222173-5GEA 270 EUR
BLDPBD222173-5G BLDPBD222173-10G
7-Bromoquinoline-2-carboxylic acid 97%
7-Bromoquinoline-2-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD11108693
CAS Number: 1057217-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR