4-(Prop-2-yn-1-yl)morpholine 97%

BD222228-5G BD222228-10G
BLDPBD222228-5GEA 27.3 EUR
BLDPBD222228-5G BLDPBD222228-10G
4-(Prop-2-yn-1-yl)morpholine 97%
4-(Prop-2-yn-1-yl)morpholine

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00510919
CAS Number: 5799-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR