2-Phenylmorpholine hydrochloride 95%
|
|
BD222302-1G
BD222302-250MG
BLDPBD222302-1GEA
92.4
EUR
BLDPBD222302-1G
BLDPBD222302-250MG
2-Phenylmorpholine hydrochloride 95%
2-Phenylmorpholine hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₀H₁₃NO·ClH Storage Temperature: Ambient |
MDL Number:
MFCD03869024 CAS Number: 23972-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...