6-Methyl-5-(trifluoromethyl)pyridin-2-amine 98%

BD222361-1G BD222361-100MG
BLDPBD222361-1GEA 401 EUR
BLDPBD222361-1G BLDPBD222361-100MG
6-Methyl-5-(trifluoromethyl)pyridin-2-amine 98%
6-Methyl-5-(trifluoromethyl)pyridin-2-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₇F₃N₂
MW: 176.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09881222
CAS Number: 1023813-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR