tert-Butyl (3,3-difluorocyclopentyl)carbamate 98%

BD222499-1G BD222499-250MG
BLDPBD222499-1GEA 786 EUR
BLDPBD222499-1G BLDPBD222499-250MG
tert-Butyl (3,3-difluorocyclopentyl)carbamate 98%
tert-Butyl (3,3-difluorocyclopentyl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₀H₁₇F₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16657108
CAS Number: 1215071-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR