1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile 96%

BD222714-100MG BD222714-5G
BLDPBD222714-100MGEA 16.8 EUR
BLDPBD222714-100MG BLDPBD222714-5G
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile 96%
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₄N₄
Storage Temperature: Refrigerator
MDL Number: MFCD13176785
CAS Number: 956010-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR