2,4-Dichloro-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine hydrochloride 98%
|
|
BD222748-5G
BD222748-100MG
BLDPBD222748-5GEA
958
EUR
BLDPBD222748-5G
BLDPBD222748-100MG
2,4-Dichloro-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine hydrochloride 98%
2,4-Dichloro-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₆Cl₃N₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD09835495 CAS Number: 1190927-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...