(2-Ethylphenyl)methanamine 95%

BD222980-100MG BD222980-250MG
BLDPBD222980-100MGEA 224 EUR
BLDPBD222980-100MG BLDPBD222980-250MG
(2-Ethylphenyl)methanamine 95%
(2-Ethylphenyl)methanamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₁₃N
Storage Temperature: Refrigerator
MDL Number: MFCD06212677
CAS Number: 53759-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR