2-Amino-5-chlorobenzophenone 98%

BD22367-1000G BD22367-25G
BLDPBD22367-1000GEA 110 EUR
BLDPBD22367-1000G BLDPBD22367-25G
2-Amino-5-chlorobenzophenone 98%
2-Amino-5-chlorobenzophenone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₃H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD00007839
CAS Number: 719-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR