tert-Butyl 3-bromopropanoate 98%

BD223944-5G BD223944-1G
BLDPBD223944-5GEA 69.3 EUR
BLDPBD223944-5G BLDPBD223944-1G
tert-Butyl 3-bromopropanoate 98%
tert-Butyl 3-bromopropanoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₁₃BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD04974087
CAS Number: 55666-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR