6-Nitro-1H-indazole 95%

BD22513-500G BD22513-100G BD22513-10G
BLDPBD22513-500GEA 189 EUR
BLDPBD22513-500G BLDPBD22513-100G BLDPBD22513-10G
6-Nitro-1H-indazole 95%
6-Nitro-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00005695
CAS Number: 7597-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR