2-Amino-5-methyl-3-nitropyridine 97%

BD22515-10G BD22515-1G
BLDPBD22515-10GEA 67.2 EUR
BLDPBD22515-10G BLDPBD22515-1G
2-Amino-5-methyl-3-nitropyridine 97%
2-Amino-5-methyl-3-nitropyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00047442
CAS Number: 7598-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR