2,2-Dimethyl-1,3-dioxan-5-amine 95%

BD225562-100MG BD225562-250MG
BLDPBD225562-100MGEA 34.7 EUR
BLDPBD225562-100MG BLDPBD225562-250MG
2,2-Dimethyl-1,3-dioxan-5-amine 95%
2,2-Dimethyl-1,3-dioxan-5-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₁₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09056824
CAS Number: 40137-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR