(4-Chlorophenyl)methanamine hydrochloride 98%

BD225931-100MG BD225931-250MG
BLDPBD225931-100MGEA 31.5 EUR
BLDPBD225931-100MG BLDPBD225931-250MG
(4-Chlorophenyl)methanamine hydrochloride 98%
(4-Chlorophenyl)methanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₉Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD00672960
CAS Number: 42365-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR