2-Methylthiazol-5-amine 95%

BD226086-1G BD226086-250MG
BLDPBD226086-1GEA 377 EUR
BLDPBD226086-1G BLDPBD226086-250MG
2-Methylthiazol-5-amine 95%
2-Methylthiazol-5-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₄H₆N₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11847087
CAS Number: 89281-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR