t-Butyl 1,3-dihydroxypropan-2-ylcarbamate 97%

BD227276-100G BD227276-10G
BLDPBD227276-100GEA 532 EUR
BLDPBD227276-100G BLDPBD227276-10G
t-Butyl 1,3-dihydroxypropan-2-ylcarbamate 97%
t-Butyl 1,3-dihydroxypropan-2-ylcarbamate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00270213
CAS Number: 125414-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR