Fmoc-N-Me-Val-OH 95%

BD22806-5G BD22806-250MG
BLDPBD22806-5GEA 27.3 EUR
BLDPBD22806-5G BLDPBD22806-250MG
Fmoc-N-Me-Val-OH 95%
Fmoc-N-Me-Val-OH

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₂₁H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00153395
CAS Number: 84000-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR