2-Chloro-6-methoxyaniline 98%

BD228128-5G BD228128-10G
BLDPBD228128-5GEA 281 EUR
BLDPBD228128-5G BLDPBD228128-10G
2-Chloro-6-methoxyaniline 98%
2-Chloro-6-methoxyaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD14455911
CAS Number: 158966-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR