H-Lys(Fmoc)-OH 97%

BD22829-100G BD22829-25G
BLDPBD22829-100GEA 365 EUR
BLDPBD22829-100G BLDPBD22829-25G
H-Lys(Fmoc)-OH 97%
H-Lys(Fmoc)-OH

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₂₁H₂₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00038365
CAS Number: 84624-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR