Methyl-1,2,4-triazole-3-carboxylate 97%

BD2283-500G BD2283-5G BD2283-25G
BLDPBD2283-500GEA 151 EUR
BLDPBD2283-500G BLDPBD2283-5G BLDPBD2283-25G
Methyl-1,2,4-triazole-3-carboxylate 97%
Methyl 1,2,4-triazole-3-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₄H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00135989
CAS Number: 4928-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR