N-Boc-5-bromo-2,3-difluoroaniline 95%

BD228729-1G BD228729-250MG
BLDPBD228729-1GEA 492 EUR
BLDPBD228729-1G BLDPBD228729-250MG
N-Boc-5-bromo-2,3-difluoroaniline 95%
N-Boc-5-bromo-2,3-difluoroaniline

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₂BrF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD12025956
CAS Number: 1150114-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR