5-Bromo-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine 98%

BD229605-250MG BD229605-1G
BLDPBD229605-250MGEA 47.3 EUR
BLDPBD229605-250MG BLDPBD229605-1G
5-Bromo-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine 98%
5-Bromo-2-(3,5-dimethyl-1H-pyrazol-1-yl)pyridine

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₀BrN₃
MW: 252.1105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026034
CAS Number: 1150164-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR