2-Methyl-4-(piperidin-1-ylsulphonyl)phenylboronic acid 96%

BD230545-100MG BD230545-5G
BLDPBD230545-100MGEA 41 EUR
BLDPBD230545-100MG BLDPBD230545-5G
2-Methyl-4-(piperidin-1-ylsulphonyl)phenylboronic acid 96%
2-Methyl-4-(piperidin-1-ylsulphonyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₁₈BNO₄S
Storage Temperature: Ambient
MDL Number: MFCD13195672
CAS Number: 1217501-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR