8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine 98%

BD232385-100G BD232385-10G
BLDPBD232385-100GEA 423 EUR
BLDPBD232385-100G BLDPBD232385-10G
8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine 98%
8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₀H₉BrN₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD22126091
CAS Number: 666816-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR