5-(Aminomethyl)-2-chloropyridine 96%
|
|
BD23284-5G
BD23284-10G
BLDPBD23284-5GEA
42
EUR
BLDPBD23284-5G
BLDPBD23284-10G
5-(Aminomethyl)-2-chloropyridine 96%
5-(Aminomethyl)-2-chloropyridine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₆H₇ClN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00673153 CAS Number: 97004-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...