8-Chloro-6-methylquinoline 98%
|
|
BD233330-1G
BD233330-100MG
BLDPBD233330-1GEA
353
EUR
BLDPBD233330-1G
BLDPBD233330-100MG
8-Chloro-6-methylquinoline 98%
8-Chloro-6-methylquinoline
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₀H₈ClN Storage Temperature: Ambient |
MDL Number:
MFCD18411263 CAS Number: 19655-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...