Cyclohexanecarboxylic acid 98%

BD23342-25G BD23342-500G
BLDPBD23342-25GEA 7.4 EUR
BLDPBD23342-25G BLDPBD23342-500G
Cyclohexanecarboxylic acid 98%
Cyclohexanecarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 232
Melting Pt: 31
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00001461
CAS Number: 98-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR