Methyl-4-bromoindole-6-carboxylate 97%

BD233697-5G BD233697-10G
BLDPBD233697-5GEA 140 EUR
BLDPBD233697-5G BLDPBD233697-10G
Methyl-4-bromoindole-6-carboxylate 97%
Methyl-4-bromoindole-6-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD03867040
CAS Number: 882679-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR