2-Propylvaleric acid 98%

BD23377-100G BD23377-25G
BLDPBD23377-100GEA 39.9 EUR
BLDPBD23377-100G BLDPBD23377-25G
2-Propylvaleric acid 98%
2-Propylvaleric acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00002672
CAS Number: 99-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR