7-Iodoimidazo[1,2-a]pyridine 95%
|
|
BD234773-1G
BD234773-250MG
BD234773-100MG
BLDPBD234773-1GEA
235
EUR
BLDPBD234773-1G
BLDPBD234773-250MG
BLDPBD234773-100MG
7-Iodoimidazo[1,2-a]pyridine 95%
7-Iodoimidazo[1,2-a]pyridine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₇H₅IN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD13177252 CAS Number: 908269-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...