4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole 95%

BD234937-5G BD234937-250MG
BLDPBD234937-5GEA 648 EUR
BLDPBD234937-5G BLDPBD234937-250MG
4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole 95%
4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₁₁BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11505042
CAS Number: 1040377-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR