4-Phenylcyclohexanol 98%

BD235685-100MG BD235685-250MG
BLDPBD235685-100MGEA 59.9 EUR
BLDPBD235685-100MG BLDPBD235685-250MG
4-Phenylcyclohexanol 98%
4-Phenylcyclohexanol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013757
CAS Number: 5437-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR