5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 98%

BD236223-5G BD236223-100MG
BLDPBD236223-5GEA 648 EUR
BLDPBD236223-5G BLDPBD236223-100MG
5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 98%
5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₃H₁₇BBrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD17926519
CAS Number: 1256360-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR